Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_177045.3 | NP_796019.1 | 12 | 1403 | |
GenBank mRNA | AK043839.1 | - | 12 | 1404 | |
AK146601.1 | - | 12 | 1422 | ||
AK173271.1 | - | 8 | 831 | ||
AK189654.1 | - | 2 | 68 | ||
AK211104.1 | - | 3 | 201 | ||
BC055068.1 | - | 10 | 1045 | ||
BC062883.1 | - | 5 | 441 | ||
BC125517.1 | - | 11 | 1293 | 40130032 | |
BC137743.1 | - | 11 | 1293 | 8860762 |