Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010882.3 | NP_035012.2 | 1 | 332 | |
GenBank mRNA | AK005078.1 | - | 1 | 290 | |
AK075729.1 | - | 1 | 290 | ||
AK075735.1 | - | 1 | 278 | ||
AK076025.1 | - | 1 | 288 | ||
AK078129.1 | - | 1 | 288 | ||
AK135444.1 | - | 1 | 288 | ||
AK149086.1 | - | 1 | 288 | ||
AU080413.1 | - | 1 | 343 | ||
BC147249.1 | - | 1 | 313 | 9056296 | |
BC147250.1 | - | 1 | 313 | 9056298 | |
M80840.1 | - | 1 | 268 |