Gene Transcripts

Gene Symbol
Fgf10
Entrez Gene ID
14165
Gene Name
fibroblast growth factor 10
Gene Aliases
AEY17, BB213776, Fgf-10, Gsfaey17
Chromosome Location
Chr. 13: 118714699 - 118792573 on Build GRCm38
UniGene ID
Mm.317323
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008002.4 NP_032028.1 3 3414
GenBank mRNA AK141633.1 - 3 2737
BC048229.1 - 3 3079 6313081

View Full Product Details