Gene Transcripts

Gene Symbol
Sh3pxd2a
Entrez Gene ID
14218
Gene Name
SH3 and PX domains 2A
Gene Aliases
2310014D11Rik, AA589508, AI256723, AI413738, C230050L11, EG329070, Fish, Gm5098, Sh3md1, TSK5, Tks5
Chromosome Location
Chr. 19: 47260174 - 47464611 on Build GRCm38
UniGene ID
Mm.127560
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164717.1 NP_001158189.1 10 1082
NM_008018.4 NP_032044.2 11 1166
GenBank mRNA AJ007012.1 - 11 1166
AK085227.1 - 4 1005
BC118022.1 - 10 918 40097448

View Full Product Details