Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001290486.1 | NP_001277415.1 | 2 | 184 | |
NM_010336.2 | NP_034466.2 | 3 | 299 | ||
NM_172989.1 | NP_766577.1 | 3 | 388 | ||
GenBank mRNA | AK030330.1 | - | 3 | 352 | |
BC025425.1 | - | 3 | 298 | 5009055 | |
BY126445.1 | - | 2 | 184 | ||
BY716867.1 | - | 3 | 347 | ||
U48235.1 | - | 2 | 149 | ||
U70622.1 | - | 3 | 340 |