Gene Transcripts

Gene Symbol
Bcl6b
Entrez Gene ID
12029
Gene Name
B cell CLL/lymphoma 6, member B
Gene Aliases
Bazf
Chromosome Location
Chr. 11: 70224127 - 70229798 on Build GRCm38
UniGene ID
Mm.12930
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007528.3 NP_031554.1 3, 4 510
GenBank mRNA AB011665.1 - 2, 3 433
AK156733.1 - 3, 4 472
BC090998.1 - 3, 4 470 6750884

View Full Product Details