Gene Transcripts

Gene Symbol
Bcap29
Entrez Gene ID
12033
Gene Name
B cell receptor associated protein 29
Gene Aliases
AW208404, Bap29
Chromosome Location
Chr. 12: 31595354 - 31634658 on Build GRCm38
UniGene ID
Mm.242379
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164090.1 NP_001157562.1 8 1285
NM_007530.3 NP_031556.1 7 1448
GenBank mRNA AK090068.1 - 8 1262
AK144754.1 - 8 1259
BC003303.1 - 7 1352 3584161
BC021661.1 - 7 1459 2646883

View Full Product Details