Gene Transcripts

Gene Symbol
MYLK
Entrez Gene ID
4638
Gene Name
myosin light chain kinase
Gene Aliases
AAT7, KRP, MLCK, MLCK1, MLCK108, MLCK210, MSTP083, MYLK1, smMLCK
Chromosome Location
Chr. 3: 123612296 - 123884302 on Build GRCh38
UniGene ID
Hs.477375
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321309.1 NP_001308238.1 32 5476
NM_053025.3 NP_444253.3 33 5684
NM_053026.3 NP_444254.3 32 5477
NM_053027.3 NP_444255.3 32 5531
NM_053028.3 NP_444256.3 31 5324
XM_011512860.2 XP_011511162.1 30 5402
XM_011512861.2 XP_011511163.1 29 5201
XM_017006469.1 XP_016861958.1 16 2960
XM_017006470.1 XP_016861959.1 15 1948
XM_017006471.1 XP_016861960.1 15 2001
XM_017006472.1 XP_016861961.1 3 294
XM_017006473.1 XP_016861962.1 3 243
GenBank mRNA AB037663.1 - 16 2760
AF069601.2 - 30 5314
AF069602.1 - 30 5365
AF069603.1 - 29 5158
AF069604.1 - 10 1340
AF096773.1 - 2 318
AF096774.1 - 2 315
AF096775.1 - 2 318
AK300610.1 - 15 1934
AK314412.1 - 2 329
AK314443.1 - 2 372
AL832211.1 - 2 3906
AY339601.1 - 30 5402
AY424269.1 - 29 5197
AY424270.1 - 30 5404
BC017811.1 - 2 339
BC064420.1 - 2 358
BC100761.2 - 2 200
BC100762.1 - 2 200
BC100763.1 - 2 197
BC107783.1 - 2 342
BC113456.1 - 2 163
BC113458.1 - 2 160
CR749364.1 - 2 5144
DB016380.1 - 2 437
U48959.2 - 31 5521
X85337.1 - 17 3014
X90870.1 - 8 1034

View Full Product Details