Gene Transcripts
- Gene Symbol
- AK8
- Entrez Gene ID
- 158067
- Gene Name
- adenylate kinase 8
- Gene Aliases
- AK 8, C9orf98, DDX31
- Chromosome Location
- Chr. 9: 132725574 - 132878811 on Build GRCh38
- UniGene ID
- Hs.421340
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001317958.1 |
NP_001304887.1
|
2, 3 |
684 |
|
NM_001317959.1 |
NP_001304888.1
|
4, 5 |
869 |
|
NM_152572.2 |
NP_689785.1
|
3, 4 |
769 |
|
XM_005272169.2 |
XP_005272226.1
|
3, 4 |
308 |
|
XM_011518277.2 |
XP_011516579.1
|
3, 4 |
308 |
|
XM_011518278.2 |
XP_011516580.1
|
2, 3 |
149 |
|
GenBank mRNA |
AK057266.1 |
- |
3, 4 |
242 |
|
AK093446.1 |
- |
2, 3 |
192 |
|
AK292186.1 |
- |
3, 4 |
254 |
|
BC034776.1 |
- |
3, 4 |
266 |
5175757 |
BC050576.1 |
- |
3, 4 |
769 |
5744517 |
View Full Product Details