Gene Transcripts

Gene Symbol
Bckdhb
Entrez Gene ID
12040
Gene Name
branched chain ketoacid dehydrogenase E1, beta polypeptide
Gene Aliases
BCKDE1B, BCKDH E1-beta
Chromosome Location
Chr. 9: 83925111 - 84124240 on Build GRCm38
UniGene ID
Mm.12819
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199195.1 NP_954665.1 6 665
GenBank mRNA AK089921.1 - 6 745
AK156264.1 - 6 744
BC064099.1 - 6 665 1246285
L16992.1 - 6 737

View Full Product Details