Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001111078.1 | NP_001104548.1 | 3 | 444 | |
NM_001111079.1 | NP_001104549.1 | 3 | 588 | ||
NM_001111080.1 | NP_001104550.1 | 3 | 444 | ||
NM_010931.3 | NP_035061.3 | 3 | 588 | ||
GenBank mRNA | AF274046.1 | - | 3 | 501 | |
AK143688.1 | - | 3 | 413 | ||
AK145376.1 | - | 3 | 410 | ||
AK145419.1 | - | 3 | 409 | ||
AK145543.1 | - | 3 | 528 | ||
AK145583.1 | - | 3 | 410 | ||
AK146951.1 | - | 3 | 561 | ||
AK147046.1 | - | 3 | 528 | ||
AK151701.1 | - | 3 | 410 | ||
AK180170.1 | - | 2 | 245 | ||
AK195382.1 | - | 2 | 241 | ||
BC022167.1 | - | 3 | 516 | 4017286 | |
D87908.1 | - | 3 | 499 |