Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001271634.1 | NP_001258563.1 | 9 | 1271 | |
NM_005444.2 | NP_005435.1 | 8 | 1175 | ||
NR_073390.1 | 7 | 1065 | |||
XM_011512138.2 | XP_011510440.1 | 9 | 1294 | ||
XM_017005248.1 | XP_016860737.1 | 8 | 1764 | ||
GenBank mRNA | AB500892.1 | - | 8 | 1047 | |
AK293281.1 | - | 9 | 1271 | ||
AK296037.1 | - | 4 | 328 | ||
AK316567.1 | - | 8 | 881 | ||
BC137455.1 | - | 8 | 897 | 9021455 | |
BC137456.1 | - | 8 | 897 | 9021456 |