Gene Transcripts
- Gene Symbol
- Thap4
- Entrez Gene ID
- 67026
- Gene Name
- THAP domain containing 4
- Gene Aliases
- 2010320B01Rik
- Chromosome Location
- Chr. 1: 93705390 - 93754838 on Build GRCm38
- UniGene ID
- Mm.272356
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_025920.3 |
NP_080196.3
|
5, 6 |
1900 |
|
GenBank mRNA |
AK008304.1 |
- |
3, 4 |
284 |
|
AK008587.1 |
- |
3, 4 |
283 |
|
AK167814.1 |
- |
5, 6 |
1660 |
|
AK209843.1 |
- |
1, 2 |
115 |
|
AK212297.1 |
- |
2, 3 |
161 |
|
BC013538.1 |
- |
4, 5 |
737 |
|
BC057963.1 |
- |
4, 5 |
1198 |
|
BC063758.1 |
- |
5, 6 |
1631 |
6334411 |
BC066042.1 |
- |
5, 6 |
1835 |
6413779 |
View Full Product Details