Gene Transcripts
- Gene Symbol
- Acbd5
- Entrez Gene ID
- 74159
- Gene Name
- acyl-Coenzyme A binding domain containing 5
- Gene Aliases
- 1300014E15Rik
- Chromosome Location
- Chr. 2: 23067700 - 23114513 on Build GRCm38
- UniGene ID
- Mm.181973
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001102436.1 |
NP_001095906.1
|
6, 7 |
696 |
|
NM_001102437.1 |
NP_001095907.1
|
6, 7 |
823 |
|
NM_001102438.1 |
NP_001095908.1
|
6, 7 |
823 |
|
NM_028793.3 |
NP_083069.1
|
6, 7 |
696 |
|
GenBank mRNA |
AK005001.2 |
- |
5, 6 |
612 |
|
AK050450.1 |
- |
6, 7 |
662 |
|
AK147839.1 |
- |
6, 7 |
823 |
|
BC035202.1 |
- |
6, 7 |
752 |
|
BC053518.1 |
- |
6, 7 |
790 |
|
BC061484.1 |
- |
5, 6 |
594 |
5355466 |
BC084584.1 |
- |
6, 7 |
628 |
5257773 |
BC090641.1 |
- |
6, 7 |
647 |
|
View Full Product Details