Gene Transcripts

Gene Symbol
C1qtnf3
Entrez Gene ID
81799
Gene Name
C1q and tumor necrosis factor related protein 3
Gene Aliases
2310005P21Rik, CORS-26, CORS26, CTRP3, Corcs, Cors
Chromosome Location
Chr. 15: 10952332 - 10980162 on Build GRCm38
UniGene ID
Mm.280158
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204134.1 NP_001191063.1 4 737
NM_030888.4 NP_112150.1 4 543
GenBank mRNA AF246265.1 - 4 520
AK029352.1 - 4 520
AK075867.1 - 4 520
AK081704.1 - 4 520
AK128898.1 - 4 543
BC019636.1 - 4 746
BC055047.1 - 4 505 6486507
BC055859.1 - 4 484 4238617
DQ002396.1 - 4 421

View Full Product Details