Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023230.2 | NP_075719.1 | 4 | 737 | |
GenBank mRNA | AF303829.1 | - | 3 | 660 | |
AK002551.1 | - | 4 | 698 | ||
AK158230.1 | - | 4 | 737 | ||
AK170413.1 | - | 4 | 708 | ||
BC003449.1 | - | 4 | 724 | 2654197 | |
BC019372.1 | - | 3 | 578 | 2646198 | |
BC053973.1 | - | 1 | 20 | ||
BC092253.1 | - | 4 | 703 | 5055396 |