Gene Transcripts

Gene Symbol
Acsbg1
Entrez Gene ID
94180
Gene Name
acyl-CoA synthetase bubblegum family member 1
Gene Aliases
BG1, Bgm, E230019G03Rik, Lpd, R75185, mKIAA0631
Chromosome Location
Chr. 9: 54604880 - 54661885 on Build GRCm38
UniGene ID
Mm.20592
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053178.2 NP_444408.1 3 607
GenBank mRNA AB050554.1 - 3 607
AK054103.1 - 3 420
AK129179.1 - 3 403
BC057322.1 - 3 403 6832261

View Full Product Details