Gene Transcripts

Gene Symbol
Naaa
Entrez Gene ID
67111
Gene Name
N-acylethanolamine acid amidase
Gene Aliases
2210023K21Rik, 3830414F09Rik, Asahl
Chromosome Location
Chr. 5: 92257660 - 92278181 on Build GRCm38
UniGene ID
Mm.28890
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163687.1 NP_001157159.1 2 402
NM_025972.4 NP_080248.2 2 408
GenBank mRNA AB162194.1 - 2 374
AK008776.1 - 2 410
AK014438.1 - 2 393
AK017141.1 - 2 387
AK133485.1 - 2 393
AK133658.1 - 2 410
AK146191.1 - 2 397
BC004572.1 - 2 378 3586255

View Full Product Details