Gene Transcripts

Gene Symbol
Kcnrg
Entrez Gene ID
328424
Gene Name
potassium channel regulator
Gene Aliases
Clld4, E030012H22Rik, Gm745
Chromosome Location
Chr. 14: 61607457 - 61612833 on Build GRCm38
UniGene ID
Mm.224464
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039105.3 NP_001034194.1 2 1705
NM_206974.1 NP_996857.1 2 1739
GenBank mRNA AK053138.1 - 2 1739

View Full Product Details