Gene Transcripts

Gene Symbol
Arl5c
Entrez Gene ID
217151
Gene Name
ADP-ribosylation factor-like 5C
Gene Aliases
Arl12, BC065791
Chromosome Location
Chr. 11: 97989578 - 97996272 on Build GRCm38
UniGene ID
Mm.277896
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207231.1 NP_997114.1 6 935
GenBank mRNA AK138099.1 - 2 1762
AK154137.1 - 6 636
BC065791.1 - 6 935 3824354

View Full Product Details