Gene Transcripts

Gene Symbol
RAD50
Entrez Gene ID
10111
Gene Name
RAD50 double strand break repair protein
Gene Aliases
NBSLD, RAD502, hRad50
Chromosome Location
Chr. 5: 132556924 - 132644621 on Build GRCh38
UniGene ID
Hs.633509
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005732.3 NP_005723.2 2, 3 622
GenBank mRNA AF057299.1 - 2, 3 542
AF057300.1 - 2, 3 542
AK290597.1 - 2, 3 564
AK307439.1 - 2, 3 297
BC062603.1 - 2, 3 539
BC073850.1 - 2, 3 539
BC108282.1 - 2, 3 559
BC136436.1 - 2, 3 514 9020425
BC140005.1 - 2, 3 616
DQ655928.2 - 2, 3 225
U63139.1 - 2, 3 609

View Full Product Details