Gene Transcripts

Gene Symbol
Wscd2
Entrez Gene ID
320916
Gene Name
WSC domain containing 2
Gene Aliases
4933413A10Rik, C530024P05Rik, Gm450
Chromosome Location
Chr. 5: 113490447 - 113490942 on Build GRCm38
UniGene ID
Mm.102783
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK049685.1 - 2653
AK173026.1 - 1 2901
BC147280.1 - 1 1984 9056352
BC147281.2 - 1 1985 9056324

View Full Product Details