Gene Transcripts

Gene Symbol
Arl4c
Entrez Gene ID
320982
Gene Name
ADP-ribosylation factor-like 4C
Gene Aliases
A630084M22Rik, Arl7, LAK
Chromosome Location
Chr. 1: 88698225 - 88702191 on Build GRCm38
UniGene ID
Mm.27968
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177305.4 NP_796279.2 1 3334
GenBank mRNA AK080743.1 - 1 3288

View Full Product Details