Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_178798.3 | NP_848913.1 | 8 | 1248 | |
GenBank mRNA | AK030762.1 | - | 9 | 1050 | |
AK046548.1 | - | 7 | 1915 | ||
AK049478.1 | - | 8 | 1248 | ||
AK053899.1 | - | 8 | 1257 | ||
AK088592.1 | - | 8 | 1165 | ||
AK129099.1 | - | 5 | 2644 | ||
AK157127.1 | - | 8 | 1387 | ||
AK157379.1 | - | 7 | 1917 | ||
AK164666.1 | - | 8 | 1249 | ||
AK165629.1 | - | 8 | 1265 | ||
AK165979.1 | - | 8 | 1267 | ||
AK191440.1 | - | 4 | 275 | ||
BC038404.1 | - | 7 | 1183 | 4949962 | |
BC042898.1 | - | 5 | 1780 |