Gene Transcripts

Gene Symbol
Syt14
Entrez Gene ID
329324
Gene Name
synaptotagmin XIV
Gene Aliases
B230320I09Rik, sytXIV
Chromosome Location
Chr. 1: 192891233 - 193035862 on Build GRCm38
UniGene ID
Mm.170401
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181546.3 NP_853524.1 7, 8 1499
GenBank mRNA AB102947.1 - 7, 8 1422

View Full Product Details