Gene Transcripts

Gene Symbol
Scd4
Entrez Gene ID
329065
Gene Name
stearoyl-coenzyme A desaturase 4
Gene Aliases
ACOD4
Chromosome Location
Chr. 19: 44333326 - 44346743 on Build GRCm38
UniGene ID
Mm.313583
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183216.3 NP_899039.2 3 527
GenBank mRNA AY430080.1 - 3 527
BC141234.1 - 3 483 9055846

View Full Product Details