Gene Transcripts

Gene Symbol
Tnip2
Entrez Gene ID
231130
Gene Name
TNFAIP3 interacting protein 2
Gene Aliases
1810020H16Rik, ABIN-2, AI428870
Chromosome Location
Chr. 5: 34496096 - 34513979 on Build GRCm38
UniGene ID
Mm.28615
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139064.2 NP_620703.1 6 1668
GenBank mRNA AJ304865.1 - 6 1669
AK160165.1 - 6 1626
BC052083.1 - 7 1714 6404610

View Full Product Details