Gene Transcripts

Gene Symbol
Fmo4
Entrez Gene ID
226564
Gene Name
flavin containing monooxygenase 4
Gene Aliases
D1Ertd532e
Chromosome Location
Chr. 1: 162793188 - 162815869 on Build GRCm38
UniGene ID
Mm.155164
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144878.1 NP_659127.1 9 1762
GenBank mRNA AF461145.1 - 9 1762

View Full Product Details