Gene Transcripts

Gene Symbol
Nrros
Entrez Gene ID
224109
Gene Name
negative regulator of reactive oxygen species
Gene Aliases
E430025L02Rik, Lrcc33, Lrrc33
Chromosome Location
Chr. 16: 32142785 - 32165589 on Build GRCm38
UniGene ID
Mm.33498
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146069.4 NP_666181.2 4 2141
GenBank mRNA AK028367.1 - 2 1093
AK042502.1 - 2 943
AK084858.1 - 4 2144
AK088770.1 - 2 862
AK128897.1 - 3 981
AK167934.1 - 3 1011
BC027411.1 - 3 991 4192329

View Full Product Details