Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001164357.1 | NP_001157829.1 | 10 | 1745 | |
NM_001164358.1 | NP_001157830.1 | 11 | 2000 | ||
NM_001290558.1 | NP_001277487.1 | 10 | 1614 | ||
NM_146118.3 | NP_666230.2 | 11 | 1781 | ||
GenBank mRNA | AK049392.1 | - | 10 | 1721 | |
AK082756.1 | - | 10 | 1702 | ||
AK122568.2 | - | 11 | 1965 | ||
AK132204.1 | - | 10 | 1699 | ||
AK204504.1 | - | 1 | 138 | ||
BC004720.1 | - | 3 | 471 | ||
BC019978.1 | - | 10 | 1553 | 4239441 | |
BC022114.1 | - | 10 | 1551 | 5098924 | |
BC037109.1 | - | 11 | 1741 | 5368306 | |
BC066998.1 | - | 11 | 1968 | 5703622 |