Gene Transcripts

Gene Symbol
Med16
Entrez Gene ID
216154
Gene Name
mediator complex subunit 16
Gene Aliases
95kDa, A630083L04, Thrap5, Trap95
Chromosome Location
Chr. 10: 79894707 - 79908941 on Build GRCm38
UniGene ID
Mm.260089
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163276.1 NP_001156748.1 2 273
NM_198107.2 NP_932775.2 2 276
GenBank mRNA AK042340.1 - 2 238
AK138125.1 - 2 260
AK149345.1 - 2 257
BC057056.1 - 2 230 6813010
BY019373.1 - 2 277

View Full Product Details