Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005908.3 | NP_005899.3 | 3 | 411 | |
XM_017008203.1 | XP_016863692.1 | 4 | 397 | ||
XM_017008204.1 | XP_016863693.1 | 3 | 366 | ||
GenBank mRNA | AB209846.1 | - | 3 | 358 | |
AK291598.1 | - | 3 | 399 | ||
AK300015.1 | - | 3 | 352 | ||
AK312827.1 | - | 3 | 366 | ||
AU132611.1 | - | 3 | 411 | ||
BC015743.1 | - | 3 | 401 | 4905535 | |
EU009130.1 | - | 3 | 311 | ||
U60337.1 | - | 3 | 398 |