Gene Transcripts

Gene Symbol
Pwp2
Entrez Gene ID
110816
Gene Name
PWP2 periodic tryptophan protein homolog (yeast)
Gene Aliases
6530411D08Rik, Pwp2h, wdp103
Chromosome Location
Chr. 10: 78170910 - 78185171 on Build GRCm38
UniGene ID
Mm.103522
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029546.2 NP_083822.1 21 3615
GenBank mRNA AB041855.1 - 21 3577
AK018333.1 - 4 1345
AK088254.1 - 21 3616
AK088983.1 - 21 3616
BC031787.1 - 10 2102
BC112325.1 - 21 3581 5355810

View Full Product Details