Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_005260641.3 | XP_005260698.1 | 3 | 670 | |
XM_005260643.3 | XP_005260700.1 | 3 | 670 | ||
XM_011529130.2 | XP_011527432.1 | 3 | 759 | ||
XM_011529134.2 | XP_011527436.1 | 3 | 759 | ||
XM_017027577.1 | XP_016883066.1 | 3 | 1106 | ||
XM_017027578.1 | XP_016883067.1 | 3 | 1111 | ||
XM_017027579.1 | XP_016883068.1 | 3 | 1112 | ||
XM_017027580.1 | XP_016883069.1 | 3 | 1112 | ||
XM_017027581.1 | XP_016883070.1 | 3 | 1109 | ||
XM_017027582.1 | XP_016883071.1 | 3 | 1122 | ||
XM_017027583.1 | XP_016883072.1 | 3 | 1106 | ||
XM_017027584.1 | XP_016883073.1 | 3 | 1106 | ||
XM_017027585.1 | XP_016883074.1 | 3 | 1103 | ||
GenBank mRNA | AK289866.1 | - | 3 | 674 | |
AK313517.1 | - | 3 | 674 | ||
BP295274.1 | - | 1 | 184 | ||
BX537992.1 | - | 3 | 675 | ||
Y10376.1 | - | 3 | 649 |