Gene Transcripts

Gene Symbol
Bend7
Entrez Gene ID
209645
Gene Name
BEN domain containing 7
Gene Aliases
1110017O21Rik, E130319B15Rik
Chromosome Location
Chr. 2: 4717831 - 4802146 on Build GRCm38
UniGene ID
Mm.149539
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001190400.1 NP_001177329.1 6 1274
NM_178663.4 NP_848778.1 6 1274
GenBank mRNA AK030428.1 - 6 961
AK042822.1 - 6 1272
AK053896.1 - 6 1276
BC056476.1 - 3 318
BC096430.1 - 6 946 5041388
BC119370.1 - 6 913 8734119

View Full Product Details