Gene Transcripts

Gene Symbol
Tchp
Entrez Gene ID
77832
Gene Name
trichoplein, keratin filament binding
Gene Aliases
A930031F18Rik, AV279199
Chromosome Location
Chr. 5: 114707765 - 114722327 on Build GRCm38
UniGene ID
Mm.239698
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029992.2 NP_084268.2 3, 4 583
GenBank mRNA AK020912.1 - 3, 4 546
AK083027.1 - 3, 4 499
AK159947.1 - 3, 4 544
BC043048.1 - 3, 4 488 5704036
BU512279.1 - 3, 4 583

View Full Product Details