Gene Transcripts

Gene Symbol
Tex13
Entrez Gene ID
83555
Gene Name
testis expressed gene 13
Gene Aliases
4933403J23Rik, Tex13b
Chromosome Location
Chr. X: 140808307 - 140813451 on Build GRCm38
UniGene ID
Mm.193025
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031381.2 NP_113558.1 2, 3 543
GenBank mRNA AF285576.1 - 2, 3 543
BC116863.1 - 2, 3 519 40126182
BC119125.1 - 2, 3 519 8733874

View Full Product Details