Gene Transcripts

Gene Symbol
Trim16
Entrez Gene ID
94092
Gene Name
tripartite motif-containing 16
Gene Aliases
9130006M08Rik, AI482483, EBBP
Chromosome Location
Chr. 11: 62820195 - 62842948 on Build GRCm38
UniGene ID
Mm.117087
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053169.2 NP_444399.2 2 823
GenBank mRNA AF449496.1 - 2 574
AK053139.1 - 2 637
BC052821.1 - 3 525 30057830

View Full Product Details