Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133973.2 | NP_598734.1 | 19 | 2717 | |
GenBank mRNA | AK156466.1 | - | 19 | 2709 | |
AK172465.1 | - | 19 | 2708 | ||
AK210898.1 | - | 1 | 330 | ||
BC023120.1 | - | 19 | 2694 | 4161858 |