Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001082476.2 | NP_001075945.1 | 4 | 404 | |
NM_001252542.1 | NP_001239471.1 | 4 | 471 | ||
GenBank mRNA | AK019617.1 | - | 4 | 401 | |
AK143124.1 | - | 4 | 444 | ||
AK169885.1 | - | 4 | 463 | ||
BC049789.1 | - | 4 | 454 | 6331033 | |
BQ771207.1 | - | 4 | 371 |