Gene Transcripts
- Gene Symbol
- Mamstr
- Entrez Gene ID
- 74490
- Gene Name
- MEF2 activating motif and SAP domain containing transcriptional regulator
- Gene Aliases
- 2810022D01Rik, 5430432N15Rik, AW743872, Mastr
- Chromosome Location
- Chr. 7: 45639200 - 45648206 on Build GRCm38
- UniGene ID
- Mm.107442
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK036109.1 |
- |
5 |
1325 |
|
AK052976.1 |
- |
2 |
2360 |
|
View Full Product Details