Gene Transcripts

Gene Symbol
Iba57
Entrez Gene ID
216792
Gene Name
IBA57, iron-sulfur cluster assembly homolog (S. cerevisiae)
Gene Aliases
4930543L23Rik, A230051G13Rik, C1orf69
Chromosome Location
Chr. 11: 59155368 - 59164591 on Build GRCm38
UniGene ID
Mm.266175
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270791.1 NP_001257720.1 2 715
GenBank mRNA AK016031.1 - 2 649


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA NM_183110.1 - 649 Chr. - - [-]

View Full Product Details