Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026271.1 | NP_080547.1 | 1 | 1482 | |
GenBank mRNA | AK003790.1 | - | 1 | 1482 | |
AK011548.1 | - | 1 | 1653 | ||
AK159648.1 | - | 1 | 1615 | ||
BC027250.1 | - | 1 | 1471 | 3603588 |