Gene Transcripts

Gene Symbol
Ufsp1
Entrez Gene ID
70240
Gene Name
UFM1-specific peptidase 1
Gene Aliases
2700038N03Rik, D5Ertd655e
Chromosome Location
Chr. 5: 137294669 - 137295664 on Build GRCm38
UniGene ID
Mm.124666
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027356.2 NP_081632.1 1 681
GenBank mRNA AK012343.1 - 1 682
BC016577.1 - 1 690 3584484
BC087958.1 - 1 702 6751787

View Full Product Details