Gene Transcripts

Gene Symbol
Acss1
Entrez Gene ID
68738
Gene Name
acyl-CoA synthetase short-chain family member 1
Gene Aliases
1110032O15Rik, AI788978, Acas2, Acas2l, AceCS2
Chromosome Location
Chr. 2: 150618111 - 150668240 on Build GRCm38
UniGene ID
Mm.7044
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080575.2 NP_542142.1 14 3548
GenBank mRNA AB046742.1 - 14 3538
AK004042.1 - 1 793
AK146568.1 - 14 3549
AK173275.1 - 14 3509
BC004097.1 - 2 1763
BC030930.1 - 10 2722 4911679
BC092278.1 - 14 3577 3710505

View Full Product Details