Gene Transcripts
- Gene Symbol
- Slc38a2
- Entrez Gene ID
- 67760
- Gene Name
- solute carrier family 38, member 2
- Gene Aliases
- 5033402L14Rik, AI316867, ATA2, SAT2, mKIAA1382
- Chromosome Location
- Chr. 15: 96687386 - 96699724 on Build GRCm38
- UniGene ID
- Mm.46754
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_175121.3 |
NP_780330.2
|
2, 3 |
502 |
|
GenBank mRNA |
AK017145.1 |
- |
2, 3 |
512 |
|
AK018767.1 |
- |
1, 2 |
197 |
|
AK030917.1 |
- |
2, 3 |
513 |
|
AK031615.1 |
- |
2, 3 |
533 |
|
AK047403.1 |
- |
2, 3 |
536 |
|
AK129342.1 |
- |
2, 3 |
463 |
|
AK164568.1 |
- |
1, 2 |
85 |
|
BC027259.1 |
- |
2, 3 |
406 |
|
BC041108.1 |
- |
2, 3 |
372 |
4038546 |
BC048178.1 |
- |
2, 3 |
502 |
6825801 |
View Full Product Details