Gene Transcripts
- Gene Symbol
- Wdr46
- Entrez Gene ID
- 57315
- Gene Name
- WD repeat domain 46
- Gene Aliases
- 2310007I04Rik, Bing4, C78447, C78559
- Chromosome Location
- Chr. 17: 33940634 - 33949695 on Build GRCm38
- UniGene ID
- Mm.2437
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_020603.2 |
NP_065628.1
|
7, 8 |
732 |
|
GenBank mRNA |
BC046977.1 |
- |
7, 8 |
732 |
|
View Full Product Details