Gene Transcripts

Gene Symbol
Noc3l
Entrez Gene ID
57753
Gene Name
nucleolar complex associated 3 homolog (S. cerevisiae)
Gene Aliases
AF233884, Fad24
Chromosome Location
Chr. 19: 38788128 - 38819270 on Build GRCm38
UniGene ID
Mm.86482
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021315.2 NP_067290.2 3 447
GenBank mRNA AB077991.1 - 3 410
AK134051.1 - 3 405
AK168944.1 - 3 448
BC031132.1 - 3 410 4954951

View Full Product Details