Gene Transcripts

Gene Symbol
Trem3
Entrez Gene ID
58218
Gene Name
triggering receptor expressed on myeloid cells 3
Gene Aliases
BB134760
Chromosome Location
Chr. 17: 48247729 - 48258848 on Build GRCm38
UniGene ID
Mm.302438
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021407.3 NP_067382.1 2 394
GenBank mRNA AF241220.1 - 2 346
AK079407.1 - 2 319
BC127035.1 - 2 329 40049608
BC127036.1 - 2 329 40049609

View Full Product Details