Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021531.6 | NP_067506.2 | 13 | 1623 | |
NM_153141.1 | NP_694781.1 | 13 | 1623 | ||
GenBank mRNA | AF117887.2 | - | 13 | 1533 | |
AK158242.1 | - | 13 | 1588 | ||
AK158757.1 | - | 13 | 1463 | ||
BC003964.1 | - | 8 | 730 | ||
BC008263.1 | - | 3 | 229 | ||
BC026427.1 | - | 3 | 189 | ||
BC036974.1 | - | 13 | 1607 | 4935077 | |
BC071267.1 | - | 14 | 1739 |